2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals
CAS: 7128-64-5 | C26H26N2O2S | 430.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ALFA14928.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7803-58-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Melting Point89-93?C
Assay from Supplier's CofA≥98.5%
Appearance (Color)White
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.
Solubility
Soluble in acetone and toluene. Insoluble in water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Fourati, M. A.; Maris, T.; Skene, W. G.; Bazuin, C G.; Prud’homme, R. E. Photophysical, Electrochemical and Crystallographic Investigations of the Fluorophore 2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene. J. Phys. Chem. B 2011, 115 (43), 12362-12369.
  2. Giani, M.; Sist, P.; Berto, D.; Serrazanetti, G. P.; Ventrella, V.; Urbani, R. The organic matrix of pelagic mucilaginous aggregates in the Tyrrhenian Sea (Mediterranean Sea). Mar. Chem. 2012, 132, 83-94.