Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
CAS: 5396-89-4 | C11H12O3 | 192.214 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14915.22100 g
Catalog number ALFA14915.22
Price (MYR)
613.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
613.00
Add to cart
Specifications
Chemical Name or MaterialBenzyl acetoacetate
CAS5396-89-4
Health Hazard 1H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
Solubility
It is soluble in alkali solutions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Laura Heys .; Christopher G. Moore.; Patrick J. Murphy. The guanidine metabolites of Ptilocaulisspiculifer and related compounds; isolation and synthesis. Chem. Soc. Rev. 2000, 29, (4), 57-67.
  2. R Chong .; PS Clezy .; AJ Liepa .; AW Nichol . The chemistry of pyrrolic compounds. VII. Synthesis of 5,5'-diformyldipyrryl-methanes. Australian Journal of Chemistry . 1969, 22, (1), 229 - 238 .