Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
CAS: 375-22-4 | C4HF7O2 | 214.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA14907.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6311-35-9
IUPAC Name6-bromopyridine-3-carboxylic acid
Molecular FormulaC6H4BrNO2
InChI KeyJDJBRMNTXORYEN-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
HPLC>=96.0 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
Solubility
Miscible with water, toluene, ether, ethanol and benzene.       
Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..   
RUO – Research Use Only
General References:
  1. Moreno-González, D.; Lara, F. J.; Jurgovská, N.; Gámiz-Gracia, L.; Garcia-Campana, A. M. Determination of aminoglycosides in honey by capillary electrophoresis tandem mass spectrometry and extraction with molecularly imprinted polymers. Anal. Chim. Acta 2015, 891, 321-328.
  2. Lv, J.; Chang, H.; Wang, Y.; Wang, M.; Xiao, J.; Zhang, Q.; Cheng, Y. Fluorination on polyethylenimine allows efficient 2D and 3D cell culture gene delivery. J. Mater. Chem. B 2015, 3 (4), 642-650.