Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Used as a nutritional supplement and stabilizer in foods.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALFA14894.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS116-17-6
IUPAC Nametris(propan-2-yl) phosphite
Molecular FormulaC9H21O3P
InChI KeySJHCUXCOGGKFAI-UHFFFAOYSA-N
SMILESCC(C)OP(OC(C)C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<25 APHA
Infrared spectrumConforms
GC>=95 %
Refractive index1.4095 to 1.4115 (20°C, 589 nm)
Appearance (Form)Clear liquid
Vitamin B12 is used as a nutritional supplement and stabilizer in foods. It is also used as a medicine and animal feed supplement. It plays an important role in the synthesis of myelin. It is also used as an antipernicious anemia factor. Further, it is used to treat cyanide poisoning and hereditary deficiency of transcobalamin II.