2-(4-Hydroxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
2-(4-Hydroxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
2-(4-Hydroxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
2-(4-Hydroxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(4-Hydroxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
CAS: 501-94-0 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFA14891.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS109-78-4
IUPAC Name3-hydroxypropanenitrile
Molecular FormulaC3H5NO
InChI KeyWSGYTJNNHPZFKR-UHFFFAOYSA-N
SMILESOCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (non-U.S. sourced material)
Assay from Supplier's CofA≥96.0% (U.S. sourced material)
Refractive Index1.4240-1.4280 @ 20°C (non-U.S. sourced material)
Appearance (Color)Clear colorless to yellow to brown or pale pink