2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ALFA14847.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7646-79-9
IUPAC Nameλ²-cobalt(2+) dichloride
Molecular FormulaCl2Co
InChI KeyGVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.1% max.
Particle Size-10 Mesh