trans-Chalcone, 97%, Thermo Scientific Chemicals
trans-Chalcone, 97%, Thermo Scientific Chemicals
trans-Chalcone, 97%, Thermo Scientific Chemicals
trans-Chalcone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Chalcone, 97%, Thermo Scientific Chemicals
CAS: 614-47-1 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA14734.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-Chalcone
CAS614-47-1
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.
Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. M Najafian.; A Ebrahim-Habibi.;N Hezareh. Trans-chalcone: a novel small molecule inhibitor of mammalian alpha-amylase. Molecular biology . 201138 (3), 1617-1620.
  2. AA Sukhorukov.; BA Zadorozhnyi. The nature of the bands in the electronic absorption spectrum of trans-chalcone. Theoretical and Experimental Chemistry. 19736 (5), 490-494.