Thiosemicarbazide, 98%, Thermo Scientific Chemicals
Thiosemicarbazide, 98%, Thermo Scientific Chemicals
Thiosemicarbazide, 98%, Thermo Scientific Chemicals
Thiosemicarbazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiosemicarbazide, 98%, Thermo Scientific Chemicals
CAS: 79-19-6 | CH5N3S | 91.132 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14630.22100 g
Catalog number ALFA14630.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1184172-35-7
IUPAC Name2,6-difluoro-4-methoxypyridine
Molecular FormulaC6H5F2NO
InChI KeyHRTFKBJLWDISRQ-UHFFFAOYSA-N
SMILESCOC1=CC(F)=NC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
Solubility
Soluble in water, ethanol, alcohols and hot methanol.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Hong, L. R.; Wang, J. P.; Zhuo, Y.; Chai, Y. Q.; Zhao, M.; Gui, G. F.; Yuan, R. A regenerable electrochemiluminescence aptasensor incorporating poly(ethylenimine) and thiosemicarbazide as dual co-reactants for signal amplification. Electrochim. Acta 2015, 186, 174-181.
  2. Karthik, N.; Sethuraman, M. G. Improved copper corrosion resistance of epoxy-functionalized hybrid sol-gel monolayers by thiosemicarbazide. Ionics 2015, 21 (5), 1477-1488.