3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
Alfa Aesar 3-Aminophthalhydrazide, 98%, Quantity: 25g, Melting Point: 325 deg.C to 330 deg.C, Molecular Weight: 177.163, Percent Purity: 98%, Beilstein: 383929, CAS: 521-31-3, EINECS Number: 208-309-4, Formula Weight: 177.16, Health Hazard 1: H315-H319-H335
Have Questions?
Catalog NumberQuantity
ALFA14597.1425 g
Catalog number ALFA14597.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2462-31-9
IUPAC Namebenzyl 2-aminoacetate hydrochloridyl
Molecular FormulaC9H11ClNO2
InChI KeyBIGFBRPKXPVZGE-UHFFFAOYSA-N
SMILES[Cl].NCC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
NMRconsistent with structure
HPLC>=96.0 %
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only
General References:
  1. Reagent for the generation of chemiluminescence.
  2. Zhang, G.; Tang, Y.; Shang, J.; Wang, Z.; Yu, H.; Du, W.; Fu, Q. Flow-injection chemiluminescence method to detect a beta2 adrenergic agonist. Luminescence 2015, 30 (1), 102-109.
  3. Sampaio, T. R.; Fonseca, A. Chemiluminescence detection in urethane-acrylate microfluidic devices. Sens. Actuators, B 2015, 213, 215-221.