trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 14755-02-3 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
100 g
Catalog number ALFA14466.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS201481-37-0
IUPAC Name(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC23H27NO5
InChI KeyLZOLWEQBVPVDPR-XOBRGWDASA-N
SMILESC[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%