trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 14755-02-3 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Catalog number ALFA14466.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7681-57-4
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.0% Na₂O₅S₂ min
Impurity contentFe 0.002%
Insoluble material0.005%
Impurity contentCl 0.05%
Trace Metals(as Pb) 0.001%