Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA14450.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS13669-42-6
IUPAC Namequinoline-3-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyRYGIHSLRMNXWCN-UHFFFAOYSA-N
SMILESO=CC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1% (non-U.S. sourced material)
Appearance (Color)White to dark cream to cream to yellow to brown
FormCrystals or powder or crystalline powder or fused solid
CommentMaterial sourced in the U.S. and in other countries
Assay (GC)≥98.0%
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.
Solubility
Soluble in water, alcohol and etherSoluble in alcohol. Slightly soluble in water, methanol and ether. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. La Nasa, J.; Degano, I.; Modugno, F.; Colombini, M. P. Industrial alkyd resins: characterization of pentaerythritol and phthalic acid esters using integrated mass spectrometry. Rapid Commun. Mass Spectrom. 2015, 29 (3), 225-237.
  2. Tummanapelli, A. K.; Vasudevan, S. Estimating successive pKa values of polyprotic acids from ab initio molecular dynamics using metadynamics: the dissociation of phthalic acid and its isomers. Phys. Chem. Chem. Phys. 2015, 17 (9), 6383-6388.