Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
CAS: 121-91-5 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
Catalog number ALFA14445.0I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5000 g
Request bulk or custom format
Chemical Identifiers
CAS23582-02-7
IUPAC Name(2-{[2-(diphenylphosphanyl)ethyl](phenyl)phosphanyl}ethyl)diphenylphosphane
Molecular FormulaC34H33P3
InChI KeyAXVOAMVQOCBPQT-UHFFFAOYSA-N
SMILESC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point126°C to 132°C
Infrared spectrumConforms
Appearance (Form)Powder and/or chunks
Elemental Analysis74.1 to 78.7 % (C)
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Zhao, S. N.; Deng, Z. P.; Zhang, Z. Y.; Huo, L. H.; Gao, S. Syntheses, structures and luminescent properties of four zinc(II) coordination polymers constructed from mixed isophthalic acid and flexible unsymmetrical bis(pyridyl) ligands. Polyhedron 2015, 102, 627-633.
  2. Acharyya, S. S.; Ghosh, S.; Bal, R. Fabrication of three dimensional (3D) hierarchical Ag/WO3 flower-like catalyst materials for the selective oxidation of m-xylene to isophthalic acid. Chem. Commun. 2015, 51 (27), 5998-6001.