Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals
CAS: 75-03-6 | C2H5I | 155.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFA14444.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS560-09-8
IUPAC Name1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
Molecular FormulaC10H16O4
InChI KeyLSPHULWDVZXLIL-UHFFFAOYNA-N
SMILESCC1(C)C(CCC1(C)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
RUO – Research Use Only
General References:
  1. Nijamudheen, A.; Datta, A. Mechanism for C-I Bond Dissociation in Iodoethane, Iodobenzene, and Iodoethene for the C-C Cross Coupling Reactions over Gold Clusters. J. Phys. Chem. C 2013, 117 (41), 21433-21440.
  2. Ljubic, I.; Matasovic, B.; Bonifacic, M. An efficient buffer-mediated control between free radical substitution and proton-coupled electron transfer: dehalogenation of iodoethane by the alpha-hydroxyethyl radical in aqueous solution. Phys. Chem. Chem. Phys. 2013, 15 (41), 18001-18011.