Copper(I) oxide, 97%, Thermo Scientific Chemicals
Copper(I) oxide, 97%, Thermo Scientific Chemicals
Copper(I) oxide, 97%, Thermo Scientific Chemicals
Copper(I) oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(I) oxide, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA14436.36500 g
Catalog number ALFA14436.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS135-20-6
IUPAC NameN-nitroso-N-phenylhydroxylamine amine
Molecular FormulaC6H9N3O2
InChI KeyNJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILESN.ON(N=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Solubility(5 % in water) Passes test
Appearance (Color)White to yellow-brown
Residue after ignition=<0.05 %
Additional infopackage usually contains small pouches
View more
It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict's Test. It is used to provide a source supplemental elemental copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict′s Test. It is used to provide a source supplemental elemental copper.
Solubility
Insoluble in water. Dissolves in concentrated ammonia.
Notes
Air and moisture sensitive. Store away from air, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Jay A. Young. Copper(I) Oxide, Cu2O.J. Chem. Educ.,2003,80(3), 257.
  2. Brian W. Robinson, et al.Quantitative preparation of sulfur dioxide, for sulfur-34/sulfur-32 analyses, from sulfides by combustion with cuprous oxide.Anal. Chem.,1975,47(7), 1179-1181.
  3. Catalyst in decarboxylation reactions of arylacetic acids: Tetrahedron, 40, 3229 (1984); and of malonic acids under mild conditions: Synthesis, 1029 (1986). For oxidative decarboxylation of carboxylic acids to give aldehydes and ketones, see: Tetrahedron Lett., 25, 3819 (1984).