4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 104-01-8 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Catalog number ALFA14399.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.02 % (Coulometric)
Stabilizer75 to 400 ppm BHT
GC>=99.85 %
Peroxides=<0.005 % (as H2O2)