4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 104-01-8 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number ALFA14399.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6921-27-3
IUPAC Name3-(prop-2-yn-1-yloxy)prop-1-yne
Molecular FormulaC6H6O
InChI KeyHRDCVMSNCBAMAM-UHFFFAOYSA-N
SMILESC#CCOCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to yellow
Assay from Supplier's CofA≥97.5%