4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 104-01-8 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number ALFA14399.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS929-06-6
IUPAC Name2-(2-aminoethoxy)ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyGIAFURWZWWWBQT-UHFFFAOYSA-N
SMILESNCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4570-1.4610 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%