5,5'-Dithiobis(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
5,5'-Dithiobis(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,5'-Dithiobis(2-nitrobenzoic acid), 99%, Thermo Scientific Chemicals
CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA14331.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6132-02-1
IUPAC Namedisodium decahydrate carbonate
Molecular FormulaCH20Na2O13
InChI KeyXYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration with HCl99.0 to 102.0 %
Chloride (Cl)=<10 ppm
Sulfate (SO4)=<30 ppm
View more
Reagent for determination of sufhydryl groups.5,5'-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for determination of sufhydryl groups.5,5′-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.
Solubility
Soluble in water and ethanol.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Reacts with free thiol groups to form a highly colored (yellow) thiolate anion: Arch. Biochem. Biophys., 82, 70 (1959); used for quantitative determination of thiols. Also used in chromogenic assay of acetylcholinesterase: Enzyme Assays, R. Eisenthal and M. J. Danson, Eds., OUP, Oxford (1992), p81.
  2. Titi, H. M.; Nandi, G.; Thakuria, R.; Goldberg, I. Coordination polymers of 5,5'-dithiobis(2-nitrobenzoic acid): Synthesis, structure and topology. Inorg. Chim. Acta 2015, 426, 55-63.
  3. Li, Z. G. Chapter Six - Quantification of Hydrogen Sulfide Concentration Using Methylene Blue and 5,5'-Dithiobis(2-Nitrobenzoic Acid) Methods in Plants. Methods Enzymol. 2015, 554, 101-110.