Magnesium hydrogen phosphate trihydrate, 97+%, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 97+%, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 97+%, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 97+%, Thermo Scientific Chemicals
CAS: 7782-75-4 | H7MgO7P | 174.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA14279.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium hydrogen phosphate trihydrate
CAS7782-75-4
Recommended StorageAmbient temperatures
Quantity250 g
EINECS Number231-823-5