5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA14244.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1609-47-8
IUPAC Namediethyl dicarbonate
Molecular FormulaC6H10O5
InChI KeyFFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILESCCOC(=O)OC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
Infrared spectrumConforms
Titration excess NaOH>=96.0 %
Refractive index1.3960 to 1.4000 (20°C, 589 nm)