5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-methoxyaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA14244.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Chloro-2-methoxyaniline
CAS95-03-4
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c