D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA14207.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Refractive index1.4210 to 1.4240 (20°C, 589 nm)
Residue after evaporation=<0.01 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
View more
Biotin is a form of vitamin B that helps the body to break down fats and carbohydrates. It is used as an alternative medicine to treat or prevent biotin deficiency, which is caused by malnutrition, rapid weight loss, long-term tube feeding and other medical conditions. It is an essential vitamin which is important for amino acid and energy metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in hot water, dimethyl sulfoxide, alcohol and benzene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Fraschetti, C.; Filippi, A.; Guarcini, L.; Steinmetz, V.; Speranza, M. Structure and Conformation of Protonated d-(+)-Biotin in the Unsolvated State. J. Phys. Chem. B 2015, 119 (20), 6198-6203.
  2. Broussard, T. C.; Pakhomova, S.; Neau, D. B.; Bonnot, R.; Waldrop, G. L. Structural Analysis of Substrate, Reaction Intermediate, and Product Binding in Haemophilus influenzae Biotin Carboxylase. Biochemistry 2015, 54 (24), 3860-3870.