1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number ALFA14140.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS3158-26-7
IUPAC Name1-isocyanatooctane
Molecular FormulaC9H17NO
InChI KeyDYQFCTCUULUMTQ-UHFFFAOYSA-N
SMILESCCCCCCCCN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless to white to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.
Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Redox indicator.
  2. Chelating ligand for the determination of Fe, Pd and V.
  3. Said, N.; Muhammad, R. S.; Qamar, A.; Zahid, H. One Pot Synthesis of 1,10-Phenanthroline-based Shape-Persistent Flouroscent Macrocycle Using Sonogashira Coupling. Lett. Org. Chem. 2015, 12 (7), 504-510.