Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA14107.30250 g
Catalog number ALFA14107.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS108-39-4
IUPAC Name3-methylphenol
Molecular FormulaC7H8O
InChI KeyRLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
GC>=98.5 %
Appearance (Color)Clear colorless to light red
Infrared spectrumConforms
Appearance (Form)Viscous liquid
View more
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
RUO – Research Use Only
General References:
  1. Pekel, O. O.; Erdik, E. Reactivity of mixed organozinc and mixed organocopper reagents: 10 Comparison of the transferability of the same group in acylation of mixed and homo halozinc diorganocuprates with benzoyl chloride. A kinetic study. J. Organomet. Chem. 2014, 751, 644-653.
  2. Cao, Q.; Guo, L.; Dong, Y.; Xie, X.; Jin, L. Autocatalytic modification of coal tar pitch using benzoyl chloride and its effect on the structure of char. Fuel Process. Technol 2015, 129, 61-66.