2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
2,4-Dimethoxyaniline, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA13943.1850 g
Catalog number ALFA13943.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dimethoxyaniline
CAS2735-04-8
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H311-H331-H373-H302
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Various 2,4-dimethoxyphenylsemicarbazones were synthesized starting from 2,4-dimethoxyaniline via a phenylcarbamate intermediate. The hydrochloride of 2,4-dimethoxyaniline was prepared by intro- ducing hydrogen chloride gas into an etheral solution and washing the precipitate with absolute diethyl ether.
Solubility
Slightly soluble in water(7g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. May be incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.
General References:
  1. Rathinasabapathy Thirumurugan,; Dharmarajan Sriram,; Amrita Saxena,; James Stablesb,; Perumal Yogeeswar. 2,4-Dimethoxyphenylsemicarbazones with anticonvulsant activity against three animal models of seizures: Synthesis and pharmacological evaluation. Bioorganic & Medicinal Chemistry. 2006, 14 (6), 3106-3112.
  2. Johanna Knospe,; Dieter Steinhilber,; Thomas Herrmann,; Hermann J. Roth.Picomole. determination of 2,4-dimethoxyanilides of prostaglandins by high-performance liquid chromatography with electrochemical detection . Journal of Chromatography A. 1988, 442 444-450.