3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA13922.1850 g
Catalog number ALFA13922.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS111-64-8
IUPAC Nameoctanoyl chloride
Molecular FormulaC8H15ClO
InChI KeyREEZZSHJLXOIHL-UHFFFAOYSA-N
SMILESCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Acidity=<1 %
Appearance (Form)Clear liquid
Color scale=<80 APHA