trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
CAS: 943-89-5 | C10H10O3 | 178.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13870.0910 g
Catalog number ALFA13870.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-4-Methoxycinnamic acid
CAS943-89-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Anton Blaakmeer,; Dick van der Wal,; André Stork,; Teris A. van Beek,; Aede de Groot,; Joop J. A. van Loon. Structure-Activity Relationship of Isolated Avenanthramide Alkaloids and Synthesized Related Compounds as Oviposition Deterrents for Pieris brassicae . J. Nat. Prod., . 1994, 57 (8), 1145-1151.
  2. Ryan G. Stabile,; Andrew P. Dicks. Two-Step Semi-Microscale Preparation of a Cinnamate Ester Sunscreen Analog. J. Chem. Educ. 2004, 81 (10), 1488.