Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
CAS: 765-30-0 | C3H7N | 57.096 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13844.0910 g
Catalog number ALFA13844.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS533-37-9
IUPAC Name5H,6H,7H-cyclopenta[b]pyridine
Molecular FormulaC8H9N
InChI KeyKRNSYSYRLQDHDK-UHFFFAOYSA-N
SMILESC1CC2=CC=CN=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5425-1.5455 @ 20?C
FormLiquid
Assay (GC)≥98.5%
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.
Solubility
Miscible with water, ethanol,ether and chloroform.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only
General References:
  1. Manakhov, A.; Nečas, D.; Čechal, J.; Pavliňák, D.; Eliáš, M.; Zajíčková, L. Deposition of stable amine coating onto polycaprolactone nanofibers by low pressure cyclopropylamine plasma polymerization. Thin Solid Films 2015, 581, 7-13.
  2. Miyamura, S.; Araki, M.; Suzuki, T.; Yamaguchi, J.; Itami, K. Stereodivergent Synthesis of Arylcyclopropylamines by Sequential C-H Borylation and Suzuki-Miyaura Coupling. Angew. Chem. Int. Ed. 2015, 54 (3), 846-851.