Palmitoyl chloride, 98%, Thermo Scientific Chemicals
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
CAS: 112-67-4 | C16H31ClO | 274.873 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13812.22100 g
Catalog number ALFA13812.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPalmitoyl chloride
CAS112-67-4
Health Hazard 1H314-H317
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
Solubility
Soluble in ether.Decomposes in water and alcohol.
Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.
RUO – Research Use Only
General References:
  1. Haixing Cheng; Siguang Wang; Jintian Yang; Yongfeng Zhou; Deyue Yan; Haixing Cheng; Siguang Wang; Jintian Yang; Yongfeng Zhou; Deyue Yan. Synthesis and self-assembly of amphiphilic hyperbranched polyglycerols modified with palmitoyl chloride.Journal of Colloid and Interface Science.2009, 337 (1), 278-284.
  2. Gowdra K. Prakash; Kittappa M. Mahadevan. Enhancing the properties of wood through chemical modification with palmitoyl chloride.Applied Surface Science.2008, 254 1751-1756.