Potassium thiocyanate, 98%, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium thiocyanate, 98%, Thermo Scientific Chemicals
CAS: 333-20-0 | CKNS | 97.176 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13731.36500 g
Catalog number ALFA13731.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS932-30-9
IUPAC Name2-(aminomethyl)phenol
Molecular FormulaC7H9NO
InChI KeyKPRZOPQOBJRYSW-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Infrared spectrumConforms
Melting point127°C to 131°C
Appearance (Form)Powder
GC>=97.5 %
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.
Solubility
Soluble in water, acetone and alcohol.
Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
RUO – Research Use Only
General References:
  1. Reaction with primary amine hydrohalide salts provides a useful synthesis of N-substituted and N,N'-disubstituted thioureas: Synthesis, 1569 (2000).
  2. Zhang, G.; Chen, B.; Guo, X.; Guo, S.; Yu, Y. Iron(II)-Promoted Synthesis of 2-Aminothiazoles via CN Bond Formation from Vinyl Azides and Potassium Thiocyanate. Adv. Synth. Catal. 2015, 357 (5), 1065-1069.
  3. Wang, S.; Wu, X.; Wang, Y.; Li, Y.; Wang, L.; Chen, Y.; Li, B. Dissolution behavior of deacetylated konjac glucomannan in aqueous potassium thiocyanate solution at low temperature. RSC Adv. 2014, 4 (42), 21918-21923.