tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
Thermo Scientific Chemicals
tert-Butyl bromide, 98+%, stab. with potassium carbonate
CAS: 507-19-7 | C4H9Br | 137.02 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13727.1425 g
Catalog number ALFA13727.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butyl Bromide
Name Notestabilized with potassium carbonate
CAS507-19-7
Health Hazard 1H225-H315-H319-H335
Health Hazard 2GHS H Statement
H225
View more
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
Solubility
Immiscible with water.
Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only