4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 99-96-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA13700.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS615-43-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or needles
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White to cream to orange to brown
4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4'-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4′-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.
Solubility
Soluble in water, alcohol, ethanol, ether, n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Froissard, D.; Rapior, S.; Bessiere, J. M.; Buatois, B.; Fruchier, A.; Sol, V.; Fons, F. Asplenioideae Species as a Reservoir of Volatile Organic Compounds with Potential Therapeutic Properties. Nat. Prod. Commun. 2015, 10 (6), 1079-1083.
  2. Albarran, G.; Esparza, M.; Mendoza, E. Oxidation of hydroxybenzoic acids by ∙OH radical produced radiolytically. Radiat. Phys. Chem. 2015, 107, 109-114.