p-Toluidine, 99+%, Thermo Scientific Chemicals
p-Toluidine, 99+%, Thermo Scientific Chemicals
p-Toluidine, 99+%, Thermo Scientific Chemicals
p-Toluidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluidine, 99+%, Thermo Scientific Chemicals
CAS: 106-49-0 | C7H9N | 107.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA13698.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Toluidine
CAS106-49-0
Health Hazard 1H301+H311+H331-H317-H319-H351
Health Hazard 2GHS H Statement
H301-H311-H331-H351-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
View more
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.
Solubility
Soluble in ethanol, pyridine, diethyl ether, acetone, carbon tetrachloride, methanol, carbon disulfide, oils and dilute acids.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Singh, K.; Dharaiya, N.; Marangoni, D. G.; Bahadur, P. Dissimilar effects of solubilized p-toluidine on the shape of micelles of differently charged surfactants. Colloids Surf., A 2015, 436, 521-529.
  2. Jiang, G.; Tang, B.; Chen, H.; Liu, Y.; Li, L.; Huang, Q.; Chen, W. Controlled growth of hexagonal Zn2GeO4 nanorods on carbon fibers for photocatalytic oxidation of p-toluidine. RSC Adv. 2015, 5 (33), 25801-25805.