n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
CAS: 1085-12-7 | C14H20O3 | 236.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA13555.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13446-24-7
IUPAC Namedimagnesium(2+) (phosphonatooxy)phosphonate
Molecular FormulaMg2O7P2
InChI KeyXZTWHWHGBBCSMX-UHFFFAOYSA-J
SMILES[Mg++].[Mg++].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)21% Mg min
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only