1,4-Bis(2-methylstyryl)benzene, 99%, Thermo Scientific Chemicals
1,4-Bis(2-methylstyryl)benzene, 99%, Thermo Scientific Chemicals
1,4-Bis(2-methylstyryl)benzene, 99%, Thermo Scientific Chemicals
1,4-Bis(2-methylstyryl)benzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Bis(2-methylstyryl)benzene, 99%, Thermo Scientific Chemicals
CAS: 13280-61-0 | C24H22 | 310.44 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13534.065 g
Catalog number ALFA13534.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.