5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13461.30250 g
Catalog number ALFA13461.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS60-35-5
IUPAC Nameacetamide
Molecular FormulaC2H5NO
InChI KeyDLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILESCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point (clear melt)76.0-83.0?C
Appearance (Color)Colorless to white
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
View more
5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([?]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([μ]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.
Solubility
Soluble in water
Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from strong bases and oxidizing agent.
RUO – Research Use Only
General References:
  1. David L Chin et. al. Rapid determination of PEGylated liposomal doxorubicin and its major metabolite in human plasma by ultraviolet-visible high-performance liquid chromatography. Journal of Chromatography B. 2002, 779(2), 259-269.
  2. Fan S-R; and Zhu L-G. Poly [[tris (di-2-pyridylamine) tris (-5-sulfonatosalicylato) tricopper (II)] trihydrate]. Acta Crystallogr. Sect. E Struct. Rep. Online, 2004, 61 (1), m174-m176.
  3. Deproteinization reagent: J. Chromat., 74, 255 (1973); 570, 285 (1991). Spray reagent for detection of sugars on TLC: J. Chromat., 315, 401 (1985).
  4. Acid catalyst, alternative to tosic acid in certain esterification and acetalization reactions: Org. Synth. Coll., 1, 246 (1941); J. Am. Chem. Soc., 76, 743 (1954); 80, 6613 (1958); J. Org. Chem., 28, 2425 (1963).