beta-Ionone, 96%, Thermo Scientific Chemicals
beta-Ionone, 96%, Thermo Scientific Chemicals
beta-Ionone, 96%, Thermo Scientific Chemicals
beta-Ionone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
beta-Ionone, 96%, Thermo Scientific Chemicals
CAS: 79-77-6 | C13H20O | 192.302 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13400.22100 g
Catalog number ALFA13400.22
Price (MYR)
315.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
315.00
Add to cart
Specifications
Chemical Name or Materialbeta-Ionone
CAS79-77-6
Melting Point-49°C
Recommended StorageAmbient temperatures
Density0.943
View more
It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.
Solubility
Soluble in water (0.11 mg/mI at 20°C).
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Jinming Zhou; Guoyan Geng; Qingwen Shi; Francoise Sauriol; Jian Hui Wu. Design and synthesis of androgen receptor antagonists with bulky side chains for overcoming antiandrogen resistance. Journal of medicinal and pharmaceutical chemistry. 2009, 52 (17), 5546-5550.
  2. Sivakumar Sekharan; Ahmet Altun; Keiji Morokuma. QM/MM study of dehydro and dihydro β-ionone retinal analogues in squid and bovine rhodopsins: implications for vision in salamander rhodopsin. Journal of the American Chemical Society, 2010, 132 (45), 15856-15859.