trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
CAS: 1013-96-3 | C9H7NO4 | 193.158 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13171.1425 g
Catalog number ALFA13171.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS927-58-2
IUPAC Name4-bromobutanoyl chloride
Molecular FormulaC4H6BrClO
InChI KeyLRTRXDSAJLSRTG-UHFFFAOYSA-N
SMILESClC(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Refractive Index1.4900-1.4950 @ 20?C