3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals
3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals
3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals
3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals
CAS: 3179-31-5 | C2H3N3S | 101.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13120.22100 g
Catalog number ALFA13120.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Mercapto-1,2,4-triazole
CAS3179-31-5
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
Solubility
Soluble in hot water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. V.Krishnakumar; R.John Xavier. FT Raman and FT-IR spectral studies of 3-mercapto-1,2,4-triazole. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2004, 60, (3),709-714
  2. H.N.Awadalla; N.A.Hammouda; N.A.Shaaban; M.Youssef; N.N.Atta; S.T El Mansoury. A histochemical study of muscles of mice infected with Trichinella spiralis. Journal of the Egyptian Society of Parasitology. 1982, 12, (1),33-39