2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 99-66-1 | C8H16O2 | 144.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12962.1425 g
Catalog number ALFA12962.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2-Di-n-propylacetic acid
CAS99-66-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H360-H314-H318-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.