Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12928.22100 g
Catalog number ALFA12928.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS84-54-8
IUPAC Name2-methyl-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC15H10O2
InChI KeyNJWGQARXZDRHCD-UHFFFAOYSA-N
SMILESCC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting Point (clear melt)171.0-180.0?C
FormPowder
Assay (GC)≥96.0%
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. H. Wu, et al. Methyl 4-nitro­benzoate.Acta Cryst.,2009,E653096.
  2. Peter. J. Groenewegen. Novel degradative pathway of 4-nitrobenzoate in Comamonas acidovorans NBA-10.Journal of’ General Microbiolog.,1992,1381599-1605.