Zinc sulfate heptahydrate, 98%, Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 98%, Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 98%, Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA12915.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS12246-51-4
IUPAC Nametetrakis(cyclooctene) diiridium dichloride
Molecular FormulaC32H56Cl2Ir2
InChI KeyCJJIQMGSHWWMCK-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ir].[Ir].C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)41.90-43.90% Carbon
Assay (unspecified)5.80-6.80% Hydrogen
Assay (unspecified)6.90-8.90% Chlorine
Zinc sulfate heptahydrate is widely used as a coagulant in the production of rayon. It finds an application in the manufacturing of calico printing, wood preservation and lithopone. It is the source of zinc which is supplied in animal feeds, fertilizers and agricultural sprays. It is utilized as electrolytes for zinc plating and surface treating agents. It is also used as a mordant in dyeing, preservative for skin and leather and in medicine as an astringent and emetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc sulfate heptahydrate is widely used as a coagulant in the production of rayon. It finds an application in the manufacturing of calico printing, wood preservation and lithopone. It is the source of zinc which is supplied in animal feeds, fertilizers and agricultural sprays. It is utilized as electrolytes for zinc plating and surface treating agents. It is also used as a mordant in dyeing, preservative for skin and leather and in medicine as an astringent and emetic.
Solubility
Soluble in water and glycerol.
Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Girija, P.; Parthiban, S.; Mojumdar, S. C. Influence of picric acid on SHG efficiency of tris (thiourea) zinc(II) sulphate (ZTS). J. Therm. Anal. Calorim. 2015, 19 (2), 953-958.
  2. Wu, X.; Yin, X.; Chen, Z.; Yu, X.; Zeng, D.; Tan, Y. Experimental determination and model simulation of the solid-liquid equilibria in the ZnSO4 -Zn(OH)2 -H2 O system. Russ. J. Phys. Chem. A 2015, 89 (6), 958-962.