Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA12905.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS363-51-9
IUPAC Name2-chloro-6-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyZJLAWMDJTMMTQB-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5480 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Notes
Stable and hygroscopic in nature.
RUO – Research Use Only
General References:
  1. Somroop, K.; Pornprasertsuk, R.; Jinawath, S. Fabrication of Y2O3 -Doped BaZrO3 Thin Films by Electrostatic Spray Deposition. Thin Solid Films 2011, 519 (19), 6408-6412.
  2. Edgar, A.; Zimmermann, J.; von Seggern, H.; Varoy, C. R. Lanthanum-stabilized europium-doped cubic barium chloride: An efficient X-ray phosphor. J. Appl. Phys. 2010, 107 (8), 083516-7.