4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 2131-55-7 | C7H4ClNS | 169.626 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12693.065 g
Catalog number ALFA12693.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS543-80-6
IUPAC Namebarium(2+) diacetate
Molecular FormulaC4H6BaO4
InChI KeyITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES[Ba++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99.0 to ≤102.0% Ba(OOCCH₃)₂
Impurity contentCa: ≤0.05%
Impurity contentFe: ≤0.001%
Impurity contentK: ≤0.003%
Impurity contentNa: ≤0.005%