4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals
4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals
4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals
4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminobenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ALFA12673.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight137.14
Unit Size
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.
Solubility
Slightly soluble in water. Soluble in methanol and diethyl ether.
Notes
Air and light sensitive. Incompatible with ferric salts and oxidizing agents. May discolor on exposure to light and air. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Liu, B.-T.; Li, P.-S. Preparation and characterization of high-refractive-index polymer/inorganic hybrid films containing TiO2 nanoparticles prepared by 4-aminobenzoic acid. Surf. Coat. Technol. 2013, 231, 301-306.
  2. Campbell, J. L.; Blanc, J. C. Y. L.; Schneider, B. B. Probing Electrospray Ionization Dynamics Using Differential Mobility Spectrometry: The Curious Case of 4-Aminobenzoic Acid. Anal. Chem. 2012, 84 (18), 7857-7864.