4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
CAS: 92-66-0 | C12H9Br | 233.108 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFA12638.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromobiphenyl
CAS92-66-0
Health Hazard 1H302-H315-H318-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
View more
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
Solubility
Insoluble in water.
Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. A Parkinson; and S Safe. Cytochrome P-450-mediated metabolism of biphenyl and the 4-halobiphenyls. Biochemical Pharmacology. 1982, 31(10), 1849-1856.
  2. Rusling JF, et al. Electrocatalytic reactions in organized assemblies: Part I. Reduction of 4-bromobiphenyl in cationic and non-ionic micelles. J. Electroanal. Chem. Interfac. Electrochem. 1988, 240(1), 201-216.