Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12619.065 g
Catalog number ALFA12619.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7790-80-9
IUPAC Namecadmium(2+) diiodide
Molecular FormulaCdI2
InChI KeyOKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES[Cd++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Total Metal Impurities0.0015% max.
Particle Size-10 Mesh
Appearance (Color)White
Residual water< 100 ppm
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.
Solubility
Soluble in water and alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Shao, X.; Ge, H.; Li, Z.; Ren, C.; Wang, J. Solubility of methylphosphonic acid in selected organic solvents. Fluid Phase Equilib. 2015, 390, 7-13.
  2. Rodin, I.; Baygildiev, T.; Stavrianidi, A.; Braun, A.; Rybalchenko, I.; Shpigun, O. Hydrophilic Interaction Liquid Chromatography Tandem Mass Spectrometry Methylphosphonic Acid Determination in Water Samples after Derivatization with p-Bromophenacyl Bromide. Chromatographia 2015, 78 (8), 585-591.