p-Toluic hydrazide, 98%, Thermo Scientific Chemicals
p-Toluic hydrazide, 98%, Thermo Scientific Chemicals
p-Toluic hydrazide, 98%, Thermo Scientific Chemicals
p-Toluic hydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluic hydrazide, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12604.1425 g
Catalog number ALFA12604.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5271-38-5
IUPAC Name2-(methylsulfanyl)ethan-1-ol
Molecular FormulaC3H8OS
InChI KeyWBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILESCSCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4934 to 1.4954 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid