3,4-Difluorophenol, 98%, Thermo Scientific Chemicals
3,4-Difluorophenol, 98%, Thermo Scientific Chemicals
3,4-Difluorophenol, 98%, Thermo Scientific Chemicals
3,4-Difluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Difluorophenol, 98%, Thermo Scientific Chemicals
CAS: 2713-33-9 | C6H4F2O | 130.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12596.065 g
Catalog number ALFA12596.06
Price (MYR)
398.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
398.00
Add to cart
Specifications
Chemical Name or Material3,4-Difluorophenol
CAS2713-33-9
Health Hazard 1H228-H302+H312+H332-H314
Health Hazard 2GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c